2-Fluoronicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

002396-5G 002396-25G 002396-100G
101783-828EA 16.92 USD
101783-828 101783-830 101783-832
2-Fluoronicotinic acid ≥98%
2-Fluoronicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD00040744
CAS Number: 393-55-5
EINECS: 206-888-8

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR