2-Acetyl-3-methylpyrazine ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(3-Methylpyrazinyl)ethan-1-one, 1-(3-Methylpyrazin-2-yl)ethanone

002387-1G 002387-5G
101783-818EA 23.08 USD
101783-818 101783-820
2-Acetyl-3-methylpyrazine ≥98%
2-Acetyl-3-methylpyrazine

Hazard code(s): Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Boiling Pt: 90 °C (1013 hPa)
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00014612
CAS Number: 23787-80-6
EINECS: 245-889-8

Order Now

Specification Test Results

Boiling point 49-50°C/1mm
Density 1.11
ND20 1.5216

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR