4-Fluoro-m-cresol ≥98% for synthesis

Supplier: Matrix Scientific
Warning

002385-5G 002385-25G
101783-814EA 27.69 USD
101783-814 101783-816
4-Fluoro-m-cresol ≥98% for synthesis
4-Fluoro-m-cresol

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₇FO
Boiling Pt: 76 °C (15mm)
Melting Pt: 32 °C
Density: 1.134 g/cm³
MDL Number: MFCD00042296
CAS Number: 452-70-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Boiling point 76°C/15mm
Melting point 32°C
Density 1.134
ND20 1.515


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR