4-Chloro-m-toluic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Chloro-3-methylbenzoic acid

002381-5G 002381-25G 002381-100G
101783-804EA 13.85 USD
101783-804 101783-806 102663-548
4-Chloro-m-toluic acid ≥97%
4-Chloro-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 216…218 °C
MDL Number: MFCD00045853
CAS Number: 7697-29-2

Order Now

Specification Test Results

Melting point 216-218°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR