2-Chloro-m-toluic acid ≥98%

Supplier: Matrix Scientific

002380-5G 002380-25G
101783-800EA 16.92 USD
101783-800 101783-802
2-Chloro-m-toluic acid ≥98%
2-Chloro-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD00079736
CAS Number: 15068-35-6

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR