4-Fluoro-3-methoxybenzonitrile ≥99%

Supplier: Matrix Scientific

002363-5G 002363-25G 002363-100G
101783-760EA 18.46 USD
101783-760 101783-762 101783-764
4-Fluoro-3-methoxybenzonitrile ≥99%
4-Fluoro-3-methoxybenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₈H₆FNO
MW: 151.14 g/mol
Boiling Pt: 96…98 °C (1.5mm)
Melting Pt: 108…109 °C
MDL Number: MFCD01569371
CAS Number: 243128-37-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 96-98°C/1.5
Melting point 108-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR