4-Fluoro-m-anisaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-3-methoxybenzaldehyde, 4-Fluoro-3-anisaldehyde

002362-5G 002362-25G 002362-100G
101783-754EA 63.08 USD
101783-754 101783-756 101783-758
4-Fluoro-m-anisaldehyde ≥97%
4-Fluoro-m-anisaldehyde

Hazard code(s): Irritant

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD00143320
CAS Number: 128495-46-5

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR