4-Fluoro-3-methoxybenzoic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Methoxy-4-fluorobenzoic acid
002361-1G
002361-5G
002361-25G
101783-748EA
13.85
USD
101783-748
101783-750
101783-752
4-Fluoro-3-methoxybenzoic acid ≥98%
4-Fluoro-3-methoxybenzoic acid
Hazard code(s): Irritant
Formula:
C₈H₇FO₃ MW: 170.14 g/mol Melting Pt: 206…208 °C |
MDL Number:
MFCD00272134 CAS Number: 82846-18-2 |
Specification Test Results
Melting point | 206-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...