4-Fluoro-3-iodotoluene ≥99%

Supplier: Matrix Scientific

002360-5G 002360-25G
101783-744EA 26.15 USD
101783-744 101783-746
4-Fluoro-3-iodotoluene ≥99%
4-Fluoro-3-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆FI
Boiling Pt: 106 °C (18mm)
MDL Number: MFCD00079759
CAS Number: 452-82-4

Order Now

Specification Test Results

Boiling point 106°C/18mm
ND20 1.77


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR