4-Chloro-3-iodobenzoic acid ≥98%

Supplier: Matrix Scientific

002355-5G 002355-25G
101783-732EA 40 USD
101783-732 101783-734
4-Chloro-3-iodobenzoic acid ≥98%
4-Chloro-3-iodobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClIO₂
Melting Pt: 222…224 °C
MDL Number: MFCD00079732
CAS Number: 42860-04-8

Order Now

Specification Test Results

Melting point 222-224°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR