4-Chloro-3-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

002344-5G 002344-25G 002344-100G
101783-704EA 21.54 USD
101783-704 101783-706 102550-818
4-Chloro-3-fluorobenzoic acid ≥97%
4-Chloro-3-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Melting Pt: 190…193 °C
MDL Number: MFCD00143290
CAS Number: 403-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR