4-Chloro-3-fluorobenzaldehyde ≥98%

Supplier: Matrix Scientific
Warning

002343-25G 002343-100G
101783-700EA 115.38 USD
101783-700 101783-702
4-Chloro-3-fluorobenzaldehyde ≥98%
4-Chloro-3-fluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄ClFO
Melting Pt: 46…49 °C
MDL Number: MFCD00143288
CAS Number: 5527-95-7

Order Now

Specification Test Results

Melting point 46-49°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR