4'-Bromo-3-chloropropiophenone ≥98%

Supplier: Matrix Scientific

002337-5G 002337-25G
101783-696EA 47.69 USD
101783-696 101783-698
4'-Bromo-3-chloropropiophenone ≥98%
4'-Bromo-3-chloropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈BrClO
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 57…62 °C
MDL Number: MFCD00018979
CAS Number: 31736-73-9
EINECS: 250-784-5

Order Now

Specification Test Results

Melting point 57-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR