4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97%
Supplier: Matrix Scientific
002330-5G
002330-25G
002330-100G
101783-686EA
60
USD
101783-686
101783-688
101783-690
4-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97%
4-Fluoro-3-(trifluoromethyl)benzaldehyde
Hazard code(s): Irritant
Formula:
C₈H₄F₄O MW: 192.11 g/mol Boiling Pt: 178 °C Density: 1.408 g/cm³ |
MDL Number:
MFCD00061314 CAS Number: 67515-60-0 |
Specification Test Results
Boiling point | 178°C |
Density | 1.408 |
ND20 | 1.457 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...