2-Chloro-3-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

002327-25G 002327-100G
101783-680EA 32.31 USD
101783-680 101783-682
2-Chloro-3-(trifluoromethyl)pyridine ≥97%
2-Chloro-3-(trifluoromethyl)pyridine

Hazard code(s): Irritant-Harmful, Irritant

Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 75 - 76 °C (30 -rr)
Melting Pt: 38 - 39 °C
MDL Number: MFCD00042223
CAS Number: 65753-47-1
EINECS: 424-520-6
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Boiling point 166-168°C
Melting point 38-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR