2-Chloro-3-(trifluoromethyl)benzaldehyde ≥97%
Supplier: Matrix Scientific
002295-1G
002295-5G
101783-626EA
60
USD
101783-626
101783-628
2-Chloro-3-(trifluoromethyl)benzaldehyde ≥97%
2-Chloro-3-(trifluoromethyl)benzaldehyde
Hazard code(s): Irritant
Formula:
C₈H₄ClF₃O MW: 208.57 g/mol |
MDL Number:
MFCD00792414 CAS Number: 93118-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...