4-Fluoro-3-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
Supplier: Matrix Scientific
Synonyms:
(4-Fluoro-3-(trifluoromethyl)phenyl)methanol
002286-1G
002286-5G
101783-614EA
30.77
USD
101783-614
101783-616
4-Fluoro-3-(trifluoromethyl)benzyl alcohol ≥97% for synthesis
4-Fluoro-3-(trifluoromethyl)benzyl alcohol
Hazard code(s): Irritant
Formula:
C₈H₆F₄O |
MDL Number:
MFCD00061171 CAS Number: 67515-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...