2-Fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: Matrix Scientific

002285-1G 002285-5G
101783-610EA 30.77 USD
101783-610 101783-612
2-Fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%
2-Fluoro-3-(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₆F₄O
MW: 194.13 g/mol
MDL Number: MFCD00061166
CAS Number: 207981-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR