4-Fluoro-3-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

002280-1G 002280-5G
101783-594EA 26.15 USD
101783-594 101783-596
4-Fluoro-3-(trifluoromethyl)benzamide ≥97%
4-Fluoro-3-(trifluoromethyl)benzamide

Hazard code(s): Irritant

Formula: C₈H₅F₄NO
Melting Pt: 84…85 °C
MDL Number: MFCD00061269
CAS Number: 67515-57-5

Order Now

Specification Test Results

Melting point 84-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR