2-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

002278-1G 002278-5G
101783-586EA 30.77 USD
101783-586 101783-588
2-Fluoro-3-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-3-(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant

Formula: C₈H₄F₄O
MW: 192.11 g/mol
Density: 1.393 g/cm³
MDL Number: MFCD00061246
CAS Number: 112641-20-0

Order Now

Specification Test Results

Density 1.393
ND20 1.455

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR