α,α,α,4-Tetrafluoro-m-toluic acid ≥97%
Supplier: Matrix Scientific
002275-25G
002275-100G
101783-578EA
73.85
USD
101783-578
101783-580
α,α,α,4-Tetrafluoro-m-toluic acid ≥97%
α,α,α,4-Tetrafluoro-m-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₄F₄O₂ |
MDL Number:
MFCD00061294 CAS Number: 67515-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...