2-Chloro-3,6-difluorobenzyl alcohol

Supplier: Matrix Scientific

002265-1G 002265-5G
101783-566EA 47.69 USD
101783-566 101783-568
2-Chloro-3,6-difluorobenzyl alcohol
2-Chloro-3,6-difluorobenzyl alcohol
Formula: C₇H₅ClF₂O
MDL Number: MFCD01631445
CAS Number: 261762-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR