4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥97%
Supplier: Matrix Scientific
002257-25G
002257-100G
101783-544EA
53.85
USD
101783-544
101783-546
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥97%
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Hazard code(s): Irritant
Formula:
C₇H₂ClF₃N₂O₄ Boiling Pt: 250 °C (1013 hPa) Melting Pt: 56…58 °C Density: 1.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007068 CAS Number: 393-75-9 EINECS: 206-889-3 UN: 2811 ADR: 6.1,II |
Specification Test Results
Melting point | 56-58°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...