5-Chloro-1,3-difluorobenzene ≥97%
Supplier: Matrix Scientific
002256-25G
002256-100G
101783-540EA
60
USD
101783-540
101783-542
5-Chloro-1,3-difluorobenzene ≥97%
5-Chloro-1,3-difluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃ClF₂ Boiling Pt: 117 °C (1013 hPa) Density: 1.329 g/cm³ (20 °C) |
MDL Number:
MFCD00041518 CAS Number: 1435-43-4 |
Specification Test Results
Boiling point | 117-119°C |
ND20 | 1.4662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...