3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,1H,2H,2H-Perfluoro-1-hexanol, 1H,1H,2H,2H-Nonafluoro-1-hexanol, 1H,1H,2H,2H-Perfluorohexan-1-ol

002225-5G 002225-25G
101783-496EA 96.92 USD
101783-496 101783-498
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol ≥97%
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol

Hazard code(s): Irritant

Formula: C₆H₅F₉O
MW: 264.09 g/mol
Boiling Pt: 140…143 °C
MDL Number: MFCD00039543
CAS Number: 2043-47-2
EINECS: 218-050-9

Order Now

Specification Test Results

Boiling point 140-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR