1H,1H,2H,2H-Perfluoro-1-iodohexane ≥98% for synthesis
Supplier: Matrix Scientific
|
Warning
|
002224-5G
002224-25G
101783-492EA
38.46
USD
101783-492
101783-494
1H,1H,2H,2H-Perfluoro-1-iodohexane ≥98% for synthesis
1H,1H,2H,2H-Perfluoro-1-iodohexane
Hazard code(s): Irritant, Refrigerate
Formula:
C₆H₄F₉I Boiling Pt: 138…140 °C Melting Pt: –25 °C Density: 1.940 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00039409 CAS Number: 2043-55-2 EINECS: 218-055-6 |
Specification Test Results
Boiling point | 138-140°C |
Melting point | -25°C |
Density | 1.94 |
ND20 | 1.374 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...