5-Chloro-2-thenoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chlorothiophene-2-carboxylic acid, 5-Chloro-2-thiophenecarboxylic acid

002208-25G 002208-100G
101783-460EA 38.46 USD
101783-460 101783-462
5-Chloro-2-thenoic acid ≥97%
5-Chloro-2-thenoic acid

Hazard code(s): Irritant

Formula: C₅H₃ClO₂S
MW: 162.6 g/mol
Melting Pt: 152…154 °C
MDL Number: MFCD00041426
CAS Number: 24065-33-6

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR