4-Fluoro-2-nitrophenol ≥99%
Supplier: Matrix Scientific
002197-25G
002197-100G
101783-454EA
27.69
USD
101783-454
101783-456
4-Fluoro-2-nitrophenol ≥99%
4-Fluoro-2-nitrophenol
Hazard code(s): Irritant
Formula:
C₆H₄FNO₃ Melting Pt: 75…77 °C |
MDL Number:
MFCD00042528 CAS Number: 394-33-2 |
Specification Test Results
Melting point | 75-77°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...