5-Fluoro-2-nitrobenzotrifluoride ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
5-Fluoro-2-nitrobenzotrifluoride, α,α,α,5-Tetrafluoro-2-nitrotoluene, 1-Fluoro-4-nitro-3-(trifluoromethyl)benzene
002195-25G
002195-100G
101783-444EA
21.54
USD
101783-444
101783-446
5-Fluoro-2-nitrobenzotrifluoride ≥98% for synthesis
5-Fluoro-2-nitrobenzotrifluoride
Hazard code(s): Irritant
Formula:
C₇H₃F₄NO₂ Boiling Pt: 198…199 °C Melting Pt: 23 °C Density: 1.49 g/cm³ (20 °C) |
MDL Number:
MFCD00039741 CAS Number: 393-09-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Boiling point | 198-199°C |
Melting point | 23°C |
Density | 1.497 |
ND20 | 1.46 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...