5-Fluoro-2-nitrobenzoic acid ≥98%

Supplier: Matrix Scientific

002192-25G 002192-100G
101783-440EA 44.62 USD
101783-440 101783-442
5-Fluoro-2-nitrobenzoic acid ≥98%
5-Fluoro-2-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄FNO₄
MDL Number: MFCD00055635
CAS Number: 320-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR