5-Fluoro-2-nitrobenzaldehyde ≥98% for synthesis
Supplier: Matrix Scientific
|
Warning
|
002191-5G
002191-25G
101783-436EA
23.08
USD
101783-436
101783-438
5-Fluoro-2-nitrobenzaldehyde ≥98% for synthesis
5-Fluoro-2-nitrobenzaldehyde
Hazard code(s): Irritant
Formula:
C₇H₄FNO₃ |
MDL Number:
MFCD00153175 CAS Number: 395-81-3 EINECS: 206-903-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...