5-Chloro-2-nitrobenzotrifluoride ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-1-nitro-2-(trifluoromethyl)benzene, 5-Chloro-2-nitrobenzotrifluoride, 1-Chloro-4-nitro-3-(trifluoromethyl)benzene, 5-Chloro-α,α,α-trifluoro-2-nitrotoluene
002190-5G
002190-25G
101783-432EA
15.38
USD
101783-432
101783-434
5-Chloro-2-nitrobenzotrifluoride ≥98%
5-Chloro-2-nitrobenzotrifluoride
Hazard code(s): Toxic
Formula:
C₇H₃ClF₃NO₂ MW: 225.55 g/mol Boiling Pt: 222…224 °C Melting Pt: 21…22 °C Density: 1.526 g/cm³ (20 °C) |
MDL Number:
MFCD00007298 CAS Number: 118-83-2 EINECS: 204-280-7 UN: 2810 ADR: 6.1,III |
Specification Test Results
Melting point | 21-22°C |
Density | 1.526 |
ND20 | 1.499 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...