3-Chloro-o-cresol ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-methylphenol, 1-Chloro-3-hydroxy-2-methylbenzene

002186-5G 002186-25G
101783-422EA 49.23 USD
101783-422 101783-424
3-Chloro-o-cresol ≥98%
3-Chloro-o-cresol

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 225 °C
Melting Pt: 91…93 °C
MDL Number: MFCD00079738
CAS Number: 3260-87-5
EINECS: 221-861-0

Order Now

Specification Test Results

Boiling point 225°C
Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR