4-Fluoro-2-methylphenol ≥97% for synthesis
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
5-Fluoro-2-hydroxytoluene, 1-Fluoro-4-hydroxy-3-methylbenzene
002178-25G
002178-100G
101783-400EA
69.23
USD
101783-400
101783-402
4-Fluoro-2-methylphenol ≥97% for synthesis
4-Fluoro-2-methylphenol
Hazard code(s): Irritant
Formula:
C₇H₇FO Boiling Pt: 84 - 85 °C (31 -rr) Melting Pt: 35 °C |
MDL Number:
MFCD00075088 CAS Number: 452-72-2 UN: 3261 ADR: 8,III |
Specification Test Results
Melting point | 37-38°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...