5-Chloro-2-methylbenzoic acid ≥98%

Supplier: Matrix Scientific

002169-5G 002169-25G
101783-370EA 18.46 USD
101783-370 101783-372
5-Chloro-2-methylbenzoic acid ≥98%
5-Chloro-2-methylbenzoic acid

Hazard code(s): Irritant

Formula: C₈H₇ClO₂
Melting Pt: 168…170 °C
MDL Number: MFCD00045842
CAS Number: 7499-06-1

Order Now

Specification Test Results

Melting point 168-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR