4-Chloro-o-toluic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
002168-5G
002168-25G
101783-364EA
18.46
USD
101783-364
101783-366
4-Chloro-o-toluic acid ≥98%
4-Chloro-o-toluic acid
Hazard code(s): Irritant
Formula:
C₈H₇ClO₂ Melting Pt: 172 °C |
MDL Number:
MFCD00045852 CAS Number: 7499-07-2 |
Specification Test Results
Melting point | 172°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...