3-Chloro-o-toluic acid ≥98%

Supplier: Matrix Scientific
Warning

002167-5G 002167-25G 002167-100G
101783-360EA 24.62 USD
101783-360 101783-362 102832-744
3-Chloro-o-toluic acid ≥98%
3-Chloro-o-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇ClO₂
Melting Pt: 164…166 °C
MDL Number: MFCD00053312
CAS Number: 7499-08-3

Order Now

Specification Test Results

Melting point 164-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR