5-Fluoro-o-toluic acid ≥99%

Supplier: Matrix Scientific
Warning

002159-5G 002159-25G 002159-100G
101783-350EA 138.46 USD
101783-350 101783-352 101783-354
5-Fluoro-o-toluic acid ≥99%
5-Fluoro-o-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 130…132 °C
MDL Number: MFCD00042294
CAS Number: 33184-16-6

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR