4-Chloro-2-methylanisole
Supplier: Matrix Scientific
Synonyms:
1-Chloro-4-methoxy-3-methylbenzene, 5-Chloro-2-methoxytoluene, 4-Chloro-1-methoxy-2-methylbenzene
002151-5G
002151-25G
101783-342EA
56.92
USD
101783-342
101783-344
4-Chloro-2-methylanisole
4-Chloro-2-methylanisole
Hazard code(s): Irritant
Formula:
C₈H₉ClO MW: 156.61 g/mol Boiling Pt: 104 °C (20mm) Melting Pt: 34…36 °C |
MDL Number:
MFCD00070771 CAS Number: 3260-85-3 EINECS: 221-860-5 |
Specification Test Results
Boiling point | 104°C/20mm |
Melting point | 34-36°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...