4-Fluoro-2-methoxyphenol

Supplier: Matrix Scientific

002149-1G 002149-5G
101783-334EA 21.54 USD
101783-334 101783-336
4-Fluoro-2-methoxyphenol
4-Fluoro-2-methoxyphenol

Hazard code(s): Irritant

Formula: C₇H₇FO₂
MW: 142.13 g/mol
Boiling Pt: 195 °C
Density: 1.25 g/cm³
MDL Number: MFCD00070797
CAS Number: 450-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR