4-Chloro-2-iodotoluene ≥98%
Supplier: Matrix Scientific
002143-25G
002143-100G
101783-318EA
55.38
USD
101783-318
101783-320
4-Chloro-2-iodotoluene ≥98%
4-Chloro-2-iodotoluene
Hazard code(s): Irritant
Formula:
C₇H₆ClI Boiling Pt: 138 °C (20mm) Density: 1.82 g/cm³ |
MDL Number:
MFCD00060661 CAS Number: 33184-48-4 |
Specification Test Results
Boiling point | 138°C/20mm |
Density | 1.82 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...