5-Chloro-2-iodotoluene ≥99%

Supplier: Matrix Scientific

002141-25G 002141-100G
101783-314EA 36.92 USD
101783-314 101783-316
5-Chloro-2-iodotoluene ≥99%
5-Chloro-2-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 118 °C (20mm)
Density: 1.82 g/cm³
MDL Number: MFCD00060665
CAS Number: 23399-70-4

Order Now

Specification Test Results

Boiling point 118°C/20mm
Density 1.82

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR