1-Fluoro-4-iodo-3-nitrobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-1-iodo-2-nitrobenzene, 5-Fluoro-2-iodonitrobenzene, 2-Iodo-5-fluoronitrobenzene

002138-5G 002138-25G
101783-306EA 32.31 USD
101783-306 101783-308
1-Fluoro-4-iodo-3-nitrobenzene ≥98%
1-Fluoro-4-iodo-3-nitrobenzene

Hazard code(s): Irritant-Harmful, Light Sensitive

Formula: C₆H₃FINO₂
MW: 267 g/mol
Boiling Pt: 162 - 163 °C (35 -rr)
Melting Pt: 22.5 °C
MDL Number: MFCD00153169
CAS Number: 364-77-2

Order Now

Specification Test Results

Boiling point 110°C/4mm
ND20 1.615

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR