1-Fluoro-2-iodobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Fluoroiodobenzene

Stabilization: Stabilised with copper

002135-25G 002135-100G
101783-302EA 38.46 USD
101783-302 101783-304
1-Fluoro-2-iodobenzene ≥99%
1-Fluoro-2-iodobenzene

(Stabilized with copper)


Hazard code(s): Irritant

Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 188…189 °C
Melting Pt: –41…–40 °C
Density: 1.903 g/cm³ (20 °C)
MDL Number: MFCD00001031
CAS Number: 348-52-7
EINECS: 206-477-3

Order Now

Specification Test Results

Boiling point 188-189°C
Melting point -41 to -40
Density 1.903
ND20 1.59

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR