5-Chloro-2-iodobenzoic acid ≥98%
Supplier: Matrix Scientific
002133-5G
002133-25G
002133-100G
101783-294EA
92.31
USD
101783-294
101783-296
101783-298
5-Chloro-2-iodobenzoic acid ≥98%
5-Chloro-2-iodobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄ClIO₂ Melting Pt: 172…174 °C |
MDL Number:
MFCD00079733 CAS Number: 13421-00-6 |
Specification Test Results
Melting point | 172-174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...