4-Chloro-2-iodobenzoic acid ≥98%
Supplier: Matrix Scientific
002132-1G
002132-5G
101783-288EA
15.38
USD
101783-288
101783-290
4-Chloro-2-iodobenzoic acid ≥98%
4-Chloro-2-iodobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄ClIO₂ MW: 282.46 g/mol Melting Pt: 163…165 °C |
MDL Number:
MFCD00060676 CAS Number: 13421-13-1 |
Specification Test Results
Melting point | 163-165°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...