5-Chloro-2-iodobenzotrifluoride ≥98%

Supplier: Matrix Scientific

002131-25G 002131-100G
101783-284EA 35.38 USD
101783-284 101783-286
5-Chloro-2-iodobenzotrifluoride ≥98%
5-Chloro-2-iodobenzotrifluoride

Hazard code(s): Irritant, Air Sensitive, Light Sensitive

Formula: C₇H₃ClF₃I
MW: 306.45 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 29…31 °C
Density: 1.97 g/cm³ (20 °C)
MDL Number: MFCD00042579
CAS Number: 23399-77-1
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR