4-Chloro-2-iodoanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-chloro-2-iodo-1-methoxybenzene

002130-5G 002130-25G
101783-280EA 21.54 USD
101783-280 101783-282
4-Chloro-2-iodoanisole ≥98%
4-Chloro-2-iodoanisole

Hazard code(s): Irritant

Formula: C₇H₆ClIO
MW: 268.48 g/mol
Boiling Pt: 105 °C (1mm)
Density: 1.84 g/cm³
MDL Number: MFCD00052858
CAS Number: 52807-27-9

Order Now

Specification Test Results

Boiling point 105°C/1mm
Density 1.84

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR