4-Chlorosalicylic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Chloro-2-hydroxybenzoic acid

002126-25G 002126-100G
101783-262EA 20 USD
101783-262 101783-264
4-Chlorosalicylic acid ≥98%
4-Chlorosalicylic acid

Hazard code(s): Irritant

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00002449
CAS Number: 5106-98-9
EINECS: 225-829-7

Order Now

Specification Test Results

Melting point 207°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR