3-Fluorosalicylic acid ≥95%

Supplier: Matrix Scientific

002125-5G 002125-25G
101783-260EA 67.69 USD
101783-260 102550-814
3-Fluorosalicylic acid ≥95%
3-Fluorosalicylic acid

Hazard code(s): Irritant

Formula: C₇H₅FO₃
MW: 156.11
MDL Number: MFCD00153167
CAS Number: 341-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR